ContactUsLinkComponent
Biological indicators of steam sterilization processes containing Geobacillus stearothermophilus (ATCC 7953) spores in suspension 1 * 1 mL
  LIOF82002
Biological indicators of steam sterilization processes containing Geobacillus stearothermophilus (ATCC 7953) spores in suspension 1 * 1 mL
  LIOF82002