CX-1 PH GRADIENT BUFFER B, PH 10.2, 1L 1 * 1 items
Catalog # 555-0139
Supplier: Thermo Fisher Scientific
CX-1 PH GRADIENT BUFFER B, PH 10.2, 1L 1 * 1 items
Catalog # 555-0139
Supplier: Thermo Fisher Scientific
Frequently Bought Together
Sodium hydroxide 98.5-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Visual colour kits, nitrite, MQuant®, Supelco®
Supplier: Merck
Chinese ink, red, Q Path® Stain_histology
Supplier: VWR Chemicals
Kjeldahl tablets, Kjeltabs
Supplier: THOMPSON & CAPPER
FIX RAL 555 2.5L 1 * 2,5 L
Supplier: RAL
Trifluoroacetic acid ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Hydrogen peroxide 6% (w/v) (20 volumes), GPR RECTAPUR®
Supplier: VWR Chemicals
Visual colour kits, chlorine, MQuant®, Supelco®
Supplier: Merck
Colorimetric rapid tests, ammonium, VISOCOLOR®
Supplier: MACHEREY-NAGEL
Methylene blue hydrate, high purity biological stain
Supplier: Thermo Fisher Scientific
BIOTECTOR 1.8N SULFURIC ACID 80MG/L MN 1 * 20 L
Supplier: Hach
Bromophenol blue sodium salt in ACS Reagent grade water 90% (dye content), Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Accessories
Supplier: Hach
NANOCOLOR® Photometric Standard Test Kits for Nitrate
Supplier: MACHEREY-NAGEL
Test kits, photometric, COD, LCK cuvette tests
Supplier: Hach
PARAPLAST REGULAR BULK PACK 56C 1 * 5 kg
Supplier: LEICA HISTOLOGY