ContactUsLinkComponent
GOMORI TRICHROME STAIN WILL STAIN MUSCLE FIBERS AND CYTOPLASM IN RED, NUCLEI IN BLACK AND COLLAGEN CONTAINING CONNECTIVE TISSUES IN BLUE. 1 * 500 mL
Catalog # GENOBAQ082
Supplier:  G-Biosciences
GOMORI TRICHROME STAIN WILL STAIN MUSCLE FIBERS AND CYTOPLASM IN RED, NUCLEI IN BLACK AND COLLAGEN CONTAINING CONNECTIVE TISSUES IN BLUE. 1 * 500 mL
Catalog # GENOBAQ082
Supplier:  G-Biosciences