ContactUsLinkComponent
Pectolyase Y-23 is a highly purified maceration enzyme from Aspergillus niger. It contains two types of pectinase such as endopolyga lacturonase and endo-pectin lyase in high activity. 1 * 1 g
Catalog # 08320971.
Supplier:  MP Biomedicals
Pectolyase Y-23 is a highly purified maceration enzyme from Aspergillus niger. It contains two types of pectinase such as endopolyga lacturonase and endo-pectin lyase in high activity. 1 * 1 g
Catalog # 08320971.
Supplier:  MP Biomedicals
Frequently Bought Together