Detect Human Cd21/CR2 with <20pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supe rnates, cell lysates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # BSBTFEK1315
Supplier: Boster Bio