ContactUsLinkComponent
Detect Human TNFRSF14 with <1pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture super nates, cell lysates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # BSBTFEK1226
Supplier:  Boster Bio
Detect Human TNFRSF14 with <1pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture super nates, cell lysates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # BSBTFEK1226
Supplier:  Boster Bio