ContactUsLinkComponent
Detect Human CD6 with <10pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernate s, cell lysates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # BSBTFEK1221
Supplier:  Boster Bio
Detect Human CD6 with <10pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernate s, cell lysates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # BSBTFEK1221
Supplier:  Boster Bio