ContactUsLinkComponent
Detect Mouse Progranulin/GRN with <10pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell cultu re supernates, cell lysates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # BSBTFEK1192
Supplier:  Boster Bio
Detect Mouse Progranulin/GRN with <10pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell cultu re supernates, cell lysates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # BSBTFEK1192
Supplier:  Boster Bio