Detect Rat E Selectin/SELE with <10pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, serum and plasma(heparin, citrate). 1 * 1 KIT
Catalog # BSBTFEK0936
Supplier: Boster Bio