Detect Mouse GDNF with <10pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernat es, cell lysates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # BSBTFEK0935
Supplier: Boster Bio