Detect Human Retinol-Binding Protein 4/RBP4 with <10pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samp les: cell culture supernates, serum, plasma(heparin , EDTA) and urine. 1 * 1 KIT
Catalog # BSBTFEK0831
Supplier: Boster Bio