ContactUsLinkComponent
Detect Human Decorin/DCN with <10pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture s upernates, cell lysates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # BSBTFEK0749
Supplier:  Boster Bio
Detect Human Decorin/DCN with <10pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture s upernates, cell lysates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # BSBTFEK0749
Supplier:  Boster Bio