ContactUsLinkComponent
Detect Mouse Adiponectin/ADIPOQ with <10pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell cu lture supernates, serum, plasma(heparin, EDTA) and urine. 1 * 1 KIT
Catalog # BSBTFEK0596
Supplier:  Boster Bio
Detect Mouse Adiponectin/ADIPOQ with <10pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell cu lture supernates, serum, plasma(heparin, EDTA) and urine. 1 * 1 KIT
Catalog # BSBTFEK0596
Supplier:  Boster Bio