ContactUsLinkComponent
Detect Mouse Resistin/RETN with <2pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, serum and plasma(heparin). 1 * 1 KIT
Catalog # BSBTFEK0582
Supplier:  Boster Bio
Detect Mouse Resistin/RETN with <2pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, serum and plasma(heparin). 1 * 1 KIT
Catalog # BSBTFEK0582
Supplier:  Boster Bio