ContactUsLinkComponent
Detect Human Resistin/RETN with <3pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, cell lysates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # BSBTFEK0581
Supplier:  Boster Bio
Detect Human Resistin/RETN with <3pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, cell lysates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # BSBTFEK0581
Supplier:  Boster Bio