ContactUsLinkComponent
Detect Mouse Vegf/VEGFA with <2pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture sup ernates, serum and plasma(heparin, EDTA, citrate). 1 * 1 KIT
  BSBTFEK0541
Detect Mouse Vegf/VEGFA with <2pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture sup ernates, serum and plasma(heparin, EDTA, citrate). 1 * 1 KIT
  BSBTFEK0541