ContactUsLinkComponent
Detect Human TIMP2 with <2pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernat es, serum, plasma(heparin , EDTA) and saliva. 1 * 1 KIT
Catalog # BSBTFEK0522
Supplier:  Boster Bio
Detect Human TIMP2 with <2pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernat es, serum, plasma(heparin , EDTA) and saliva. 1 * 1 KIT
Catalog # BSBTFEK0522
Supplier:  Boster Bio