ContactUsLinkComponent
Detect Human P Selectin/SELP with <5pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell cultur e supernates, serum and plasma(heparin, EDTA, citrate). 1 * 1 KIT
Catalog # BSBTFEK0505
Supplier:  Boster Bio
Detect Human P Selectin/SELP with <5pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell cultur e supernates, serum and plasma(heparin, EDTA, citrate). 1 * 1 KIT
Catalog # BSBTFEK0505
Supplier:  Boster Bio