ContactUsLinkComponent
Detect Mouse L Selectin/SELL with <5pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell cultur e supernates, cell lysates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # BSBTFEK0504
Supplier:  Boster Bio
Detect Mouse L Selectin/SELL with <5pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell cultur e supernates, cell lysates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # BSBTFEK0504
Supplier:  Boster Bio