Detect Human Rantes/CCL5 with <3pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture su pernates, serum, plasma(heparin, EDTA, citrate) and urine. 1 * 1 KIT
Catalog # BSBTFEK0494
Supplier: Boster Bio