ContactUsLinkComponent
Detect Human Il-2 R Alpha/IL2RA with <5pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell cul ture supernates, cell lysates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # BSBTFEK0400
Supplier:  Boster Bio
Detect Human Il-2 R Alpha/IL2RA with <5pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell cul ture supernates, cell lysates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # BSBTFEK0400
Supplier:  Boster Bio