ContactUsLinkComponent
Detect Mouse IL2 with <1pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates , serum and plasma(heparin, EDTA, citrate). 1 * 1 KIT
Catalog # BSBTFEK0398
Supplier:  Boster Bio
Detect Mouse IL2 with <1pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates , serum and plasma(heparin, EDTA, citrate). 1 * 1 KIT
Catalog # BSBTFEK0398
Supplier:  Boster Bio