Detect Mouse Gm-Csf/CSF2 with <1pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture su pernates, cell lysates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # BSBTFEK0365
Supplier: Boster Bio