ContactUsLinkComponent
Detect Human Gm-Csf/CSF2 with <1pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture su pernates, serum and plasma(heparin, EDTA, citrate). 1 * 1 KIT
Catalog # BSBTFEK0364
Supplier:  Boster Bio
Detect Human Gm-Csf/CSF2 with <1pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture su pernates, serum and plasma(heparin, EDTA, citrate). 1 * 1 KIT
Catalog # BSBTFEK0364
Supplier:  Boster Bio