ContactUsLinkComponent
Detect Rat GDNF with <4pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, cell lysates, serum and plasma (heparin, EDTA). 1 * 1 KIT
  BSBTFEK0363
Detect Rat GDNF with <4pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, cell lysates, serum and plasma (heparin, EDTA). 1 * 1 KIT
  BSBTFEK0363