ContactUsLinkComponent
KGF (keratinocyte growth factor), also known as FGF-7 (fibroblast growth factor-7), is one of 22 known members. The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. 1 * 100 µG
Catalog # BSBTR01931-1
Supplier:  Boster Bio
KGF (keratinocyte growth factor), also known as FGF-7 (fibroblast growth factor-7), is one of 22 known members. The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. 1 * 100 µG
Catalog # BSBTR01931-1
Supplier:  Boster Bio