ContactUsLinkComponent
Microtitre Plates 384 Deep-well 400µl Polypropylene Square well with Round-Bottom Non-Sterile Alpha-numeric Index SBS Footprint 1 * 6 items
Catalog # ALPCLW9602
Supplier:  Alpha Laboratories
Microtitre Plates 384 Deep-well 400µl Polypropylene Square well with Round-Bottom Non-Sterile Alpha-numeric Index SBS Footprint 1 * 6 items
Catalog # ALPCLW9602
Supplier:  Alpha Laboratories
Frequently Bought Together