Ground Joint Thermometer, enclosed scale, range -10+150 in 1C, blue liquid, built-in length encl. Ground joint 14/23 68 mm, 1 * 1 items
Catalog # SCNL1302210/03/3P
Supplier: Ludwig Schneider GmbH & CO. KG