ContactUsLinkComponent
Microslide green 90 ground edge alu unimark made of soda lime glass of 3hydrolytic class in compliance with din iso 8037/1,dimensions: approx. 76 x 26 mm, thickness approx. 1 mm. 1 * 2.500 items
Catalog # MARI0704207
Supplier:  MARIENFELD
Microslide green 90 ground edge alu unimark made of soda lime glass of 3hydrolytic class in compliance with din iso 8037/1,dimensions: approx. 76 x 26 mm, thickness approx. 1 mm. 1 * 2.500 items
Catalog # MARI0704207
Supplier:  MARIENFELD