Vented Qtray with cover & 48 Well Divider, polystyrene, 242 x 240 x 20 mm 20 per case 1 * 20 items
Catalog # MLDVX6029
Supplier: MOLECULAR DEVICES
Vented Qtray with cover & 48 Well Divider, polystyrene, 242 x 240 x 20 mm 20 per case 1 * 20 items
Catalog # MLDVX6029
Supplier: MOLECULAR DEVICES
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Test strips for peroxide, MQuant®, Supelco®
Supplier: Merck
Accessories for PIPETBOY acu / pro
Supplier: Integra
VWR® Facial Tissues
Supplier: VWR Collection
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
VWR®, Nitrile Gloves
Supplier: VWR Collection
Wipes for delicate tasks, Kimtech™ Science
Supplier: KIMBERLY CLARK
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Bench Protectors
Supplier: VWR Collection
Accessories for Mettler balances
Supplier: Mettler - Toledo
VWR®, Snap Caps for Snap Ring Vials ND11
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
BUFFER PH 4,01 DOSING BOTTLE 1 * 500 mL
Supplier: HAMILTON BONADUZ
Plastic cuvette for LICO
Supplier: Hach