ContactUsLinkComponent
Clear 2-Dram Glass Screw Thread Sample Vials with Unattached 15-425 Polyvinyl-Faced Pulp Lined Black Phenolic Cap 1 * 1.728 items
Catalog # SCERSP60910C-2
Supplier:  DWK Life Sciences
Clear 2-Dram Glass Screw Thread Sample Vials with Unattached 15-425 Polyvinyl-Faced Pulp Lined Black Phenolic Cap 1 * 1.728 items
Catalog # SCERSP60910C-2
Supplier:  DWK Life Sciences
Frequently Bought Together