ContactUsLinkComponent
Connecting kit for gas filling, Connecting kit for gas filling, consisting of two discs with hose connectors of PP natural, 5 m PVC hose for gas filling ID 9 mm and 5 m PE aeration tubing ID 45 mm, including hexagon wrench and fastening screws. 1 * 1 items
Catalog # BOHLV1785-07
Supplier:  Bohlender
undefined
Connecting kit for gas filling, Connecting kit for gas filling, consisting of two discs with hose connectors of PP natural, 5 m PVC hose for gas filling ID 9 mm and 5 m PE aeration tubing ID 45 mm, including hexagon wrench and fastening screws. 1 * 1 items
Catalog # BOHLV1785-07
Supplier:  Bohlender
Frequently Bought Together