ContactUsLinkComponent
Glove Box Duo 1 with Transfer Chambers, Aluminium frame with panels made of acrylic glass, base panel made of polyethylene. Front si de with four glove ports and two pairs of natural rubber gloves size 9, three cable lead-ins on the back side (upper 1 * 1 items
Catalog # BOHLV1730-08
Supplier:  Bohlender
undefined
Glove Box Duo 1 with Transfer Chambers, Aluminium frame with panels made of acrylic glass, base panel made of polyethylene. Front si de with four glove ports and two pairs of natural rubber gloves size 9, three cable lead-ins on the back side (upper 1 * 1 items
Catalog # BOHLV1730-08
Supplier:  Bohlender
Frequently Bought Together