ContactUsLinkComponent
Filtering Rods, Made of porous PTFE, roundly machined cylinder with diameter approx. 30 mm and height approx. 115 mm. Round shape fo r better treatment. 1 * 1 items
Catalog # BOHLN1530-20
Supplier:  Bohlender
undefined
Filtering Rods, Made of porous PTFE, roundly machined cylinder with diameter approx. 30 mm and height approx. 115 mm. Round shape fo r better treatment. 1 * 1 items
Catalog # BOHLN1530-20
Supplier:  Bohlender