Glove Box Duo 1 with Transfer Chambers Antistatic, Aluminium frame with static dissipative panels made of polycarbonate, base panel made of static dissipative polyethylene. Front side with four glove ports and two pairs of static dissipative EPDM glo 1 * 1 items
Catalog # BOHLV1735-08
Supplier: Bohlender