ContactUsLinkComponent
Screw Caps with Aperture EX, Screw cap with handy knurl, suitable for glthreads, with aperture, made of conductive PPS-EX 1 * 1 items
Catalog # BOHLH898-45
Supplier:  Bohlender
undefined
Screw Caps with Aperture EX, Screw cap with handy knurl, suitable for glthreads, with aperture, made of conductive PPS-EX 1 * 1 items
Catalog # BOHLH898-45
Supplier:  Bohlender
Frequently Bought Together