Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Clamp Screws, Suitable for all BOLA Stirrer Blades for shaft O. D. 16 mm 1 * 5 items
Catalog # BOHLC950-03
Supplier:
Bohlender
Expand image
Clamp Screws, Suitable for all BOLA Stirrer Blades for shaft O. D. 16 mm 1 * 5 items
Catalog # BOHLC950-03
Supplier:
Bohlender
Frequently Bought Together
VWR®, Funnels with Short Stem
Supplier: VWR Collection
Screw caps for laboratory bottles, DURAN®
Supplier: DWK Life Sciences
VWR®, Erlenmeyer Flasks
Supplier: VWR Collection
Laboratory bottles, narrow neck, with screw cap, DURAN®
Supplier: DWK Life Sciences
VWR®, Beakers, Tall Form
Supplier: VWR Collection
VWR®, Beakers, Low Form, Heavy-Duty
Supplier: VWR Collection
Screw caps for laboratory bottles, DURAN®
Supplier: DWK Life Sciences
Sodium carbonate ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
VWR®, Laboratory Bottles, Narrow Neck, with Screw Cap
Supplier: VWR Collection
VWR®, Pasteur Pipettes
Supplier: VWR Collection
VWR®, Beakers, Low Form
Supplier: VWR Collection
Macro Cells, Quartz Glass for Absorption Measurements
Supplier: HELLMA