ContactUsLinkComponent
Driving Bell G-MRK, Suitable for BOLA Magnetic Stirrer Heads G-MRK with Ground Joint ns29/32 or ns45/40 for shaft O. D. 8 mm, Cat. N o. C 502-08 and C 504-08 and BOLA Magnetic Stirrer Heads G-MRK with Ground Joint ns29/32 or ns45/40 for shaft O. D. 1 1 * 1 items
Catalog # BOHLC932-02
Supplier:  Bohlender
undefined
Driving Bell G-MRK, Suitable for BOLA Magnetic Stirrer Heads G-MRK with Ground Joint ns29/32 or ns45/40 for shaft O. D. 8 mm, Cat. N o. C 502-08 and C 504-08 and BOLA Magnetic Stirrer Heads G-MRK with Ground Joint ns29/32 or ns45/40 for shaft O. D. 1 1 * 1 items
Catalog # BOHLC932-02
Supplier:  Bohlender