ContactUsLinkComponent
Mini-Propeller-Stirrer Shaft, PTFE-jacketed stainless steel shaft, propeller completely made of PTFE with three 45 angled angular b lades. Universal chemical resistance since the product is only exposed to PTFE. 1 * 1 items
Catalog # BOHLC482-20
Supplier:  Bohlender
undefined
Mini-Propeller-Stirrer Shaft, PTFE-jacketed stainless steel shaft, propeller completely made of PTFE with three 45 angled angular b lades. Universal chemical resistance since the product is only exposed to PTFE. 1 * 1 items
Catalog # BOHLC482-20
Supplier:  Bohlender
Frequently Bought Together