Distributor with Stirrer Bearing, Completely made of PTFE. With ground joint ns45/40 and three GL-threaded necks. Center neck serves as stirrer bearing. A special gasket made of PTFE and an FPM o-ring which is compressed by a glscrew cap provide a g 1 * 1 items
Catalog # BOHLC435-08
Supplier: Bohlender