ContactUsLinkComponent
Replacement Screw Caps EX, Screw cap with GL-thread made of PPS-EX. For sealing the stirrer shaft in BOLA Stirrer Bearings EX. Screw caps compress the o-ring of the special gasket and provide sealing of the stirrer shaft. 1 * 1 items
Catalog # BOHLC433-08
Supplier:  Bohlender
undefined
Replacement Screw Caps EX, Screw cap with GL-thread made of PPS-EX. For sealing the stirrer shaft in BOLA Stirrer Bearings EX. Screw caps compress the o-ring of the special gasket and provide sealing of the stirrer shaft. 1 * 1 items
Catalog # BOHLC433-08
Supplier:  Bohlender