ContactUsLinkComponent
U-Shaped Stirrer Shafts, PTFE-jacketed stainless steel shaft, u-shaped stirrer blade completely made of PTFE. Universal chemical res istance since the product is only exposed to PTFE. 1 * 1 items
Catalog # BOHLC384-74
Supplier:  Bohlender
undefined
U-Shaped Stirrer Shafts, PTFE-jacketed stainless steel shaft, u-shaped stirrer blade completely made of PTFE. Universal chemical res istance since the product is only exposed to PTFE. 1 * 1 items
Catalog # BOHLC384-74
Supplier:  Bohlender
Frequently Bought Together