ContactUsLinkComponent
Screw In Stopper EX, Made of PTFE-EX. For connection to BOLA Reactor Lids. For closure of non-used connectors. With hexagonal grippi ng surface in standard wrench size. Earthing is made via a connection on the Reactor Lid. 1 * 1 items
Catalog # BOHLB173-02
Supplier:  Bohlender
undefined
Screw In Stopper EX, Made of PTFE-EX. For connection to BOLA Reactor Lids. For closure of non-used connectors. With hexagonal grippi ng surface in standard wrench size. Earthing is made via a connection on the Reactor Lid. 1 * 1 items
Catalog # BOHLB173-02
Supplier:  Bohlender