Screw In Fittings glEX, Made of PTFE-EX. For connection to BOLA Reactor Lids as glnecks. With hexagonal gripping surface in standard wrench size. Earthing is made via a connection on the reactor lid. 1 * 1 items
Catalog # BOHLB172-16
Supplier: Bohlender
Frequently Bought Together