ViewPlate-96 PDL Coated, Black, Glass Bottom, Tissue Culture Treated, Sterile, 96-Well with Lid, Case of 8 1 * 8 items
PERK6005530
ViewPlate-96 PDL Coated, Black, Glass Bottom, Tissue Culture Treated, Sterile, 96-Well with Lid, Case of 8 1 * 8 items
PERK6005530
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection