ContactUsLinkComponent
EC HPLC column EC 150/4 NUCLEOSIL 120-3 C18 length: 150 mm, ID: 4 mm 1 * 1 items
Catalog # MANA720740.40
Supplier:  MACHEREY-NAGEL
EC HPLC column EC 150/4 NUCLEOSIL 120-3 C18 length: 150 mm, ID: 4 mm 1 * 1 items
Catalog # MANA720740.40
Supplier:  MACHEREY-NAGEL