ContactUsLinkComponent
Rabbit monoclonal antibody raised against synthetic phosphopeptide of human LYN. 1 * 100 µG
Catalog # ABNOMAB20536
Supplier:  Abnova
Rabbit monoclonal antibody raised against synthetic phosphopeptide of human LYN. 1 * 100 µG
Catalog # ABNOMAB20536
Supplier:  Abnova