0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
ALPINE Test Sieve 200 LS with stainless steel frame 200x25 mm, w / mm -0.038 1 * 1 items
Catalog # HAVE206160096
Supplier:
NEXOPART
Expand image
ALPINE Test Sieve 200 LS with stainless steel frame 200x25 mm, w / mm -0.038 1 * 1 items
Catalog # HAVE206160096
Supplier:
NEXOPART
Frequently Bought Together
Dishes, disposable
Supplier: BOCHEM
Spoon spatulas, analyse type
Supplier: BOCHEM
Stabilising rings for erlenmeyer
Supplier: BOCHEM
Laboratory scissors, straight
Supplier: RSG
VWR®, Magnetic Stirring Bars, Cross-Shaped
Supplier: VWR Collection
Funnel with grip and air groove
Supplier: RSG
VWR®, Magnetic stirring bars, cylindrical
Supplier: VWR Collection
VWR®, Clamps
Supplier: VWR Collection
Double spatulas, micro
Supplier: BOCHEM
VWR®, Test Sieves, 200×50 mm
Supplier: VWR Collection
Bottles, aluminium, with screw cap
Supplier: BURKLE
VWR®, Disposable Weighing and Drying Dishes or Pans
Supplier: VWR Collection
VWR®, Aluminium foil in dispensers
Supplier: VWR Collection
Chemical spoon with spatula end
Supplier: RSG
VWR®, Disposable Weighing Dishes
Supplier: VWR Collection
Test sieves
Supplier: NEXOPART