ContactUsLinkComponent
Plant Growth Chamber with programmable temperature & illumination. Temperature 0C to 50C lights off. 10C to 50C lights on.29 4 li tres . Interior capacity, supplied with 5 shelves. 1 * 1 items
Catalog # SANYMLR-352-PE
Supplier:  PHC Corporation
Plant Growth Chamber with programmable temperature & illumination. Temperature 0C to 50C lights off. 10C to 50C lights on.29 4 li tres . Interior capacity, supplied with 5 shelves. 1 * 1 items
Catalog # SANYMLR-352-PE
Supplier:  PHC Corporation
Frequently Bought Together