Caps for storage vials, ND15 PP, sealing pad, closed, Butyl/PTFE 1 * 100 items
Catalog # ROTHXC44.1
Supplier: Roth Carl
Caps for storage vials, ND15 PP, sealing pad, closed, Butyl/PTFE 1 * 100 items
Catalog # ROTHXC44.1
Supplier: Roth Carl
Frequently Bought Together
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Storage Boxes for Sample Vials and Bottles, PP
Supplier: VWR Collection
VWR®, Manual Crimper and Decapper
Supplier: VWR Collection
Exhaust filters, V3.0
Supplier: S.C.A.T.
VWR®, Snap Ring Vials ND11
Supplier: VWR Collection
Headspace crimp caps with 20 mm septa
Supplier: Agilent
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Screw Neck Headspace Vials ND18
Supplier: VWR Collection
VWR®, TLC Aluminium Plates, Silica, Unmodified Layers
Supplier: VWR Collection
Silica gel, Pore size: 60 Å 0.063-0.200 mm (70-230 mesh ASTM) for column chromatography, Millipore®
Supplier: MERCK PRODUCTION CHEMICALS
VWR®, Inserts for Short Thread Vials ND9
Supplier: VWR Collection
Certified headspace crimp top vials
Supplier: Agilent